Anti-AIPL1 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A69745-100UL A69745-50UL
77841-184EA 330 USD
77841-184 77841-185
Anti-AIPL1 Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: AIPL1
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR