Anti-RPS6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A33861-100UG A33861-50UG
77798-186EA 330 USD
77798-186 77798-187
Anti-RPS6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: RPS6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR