Anti-RFC4 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A33086-100UG A33086-50UG
77797-081EA 330 USD
77797-081 77797-082
Anti-RFC4 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RFC4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR