Anti-SCLY Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A34225-100UG A34225-50UG
77798-793EA 330 USD
77798-793 77798-794
Anti-SCLY Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SCLY
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR