Anti-C5orf22 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A56575-100UG A56575-50UG
77826-539EA 330 USD
77826-539 77826-540
Anti-C5orf22 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: C5orf22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR