Anti-SELENOP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53585-100UL A53585-50UL
77820-963EA 330 USD
77820-963 77820-964
Anti-SELENOP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: SELENOP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR