Anti-TXNDC9 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A37763-100UG A37763-50UG
77811-754EA 330 USD
77811-754 77803-754
Anti-TXNDC9 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TXNDC9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR