Anti-ompT Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A51402-100UG A51402-50UG
77817-112EA 330 USD
77817-112 77817-113
Anti-ompT Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ompT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR