Anti-CAPS2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66624-100UG A66624-50UG
77835-312EA 330 USD
77835-312 77835-313
Anti-CAPS2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CAPS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR