Anti-TKT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A37656-100UL A37656-50UL
77811-665EA 330 USD
77811-665 77803-665
Anti-TKT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: TKT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR