Anti-MS4A7 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A68757-100UG A68757-50UG
77839-325EA 330 USD
77839-325 77839-326
Anti-MS4A7 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MS4A7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR