Anti-X Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A52660-100UG A52660-50UG
77819-378EA 330 USD
77819-378 77819-379
Anti-X Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hepatitis B virus genotype D
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR