Anti-ZMPSTE24 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A38705-100UG A38705-50UG
77804-981EA 330 USD
77804-981 77804-982
Anti-ZMPSTE24 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: ZMPSTE24
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR