Anti-PPP1R3A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A54318-100UL A54318-50UL
77822-267EA 330 USD
77822-267 77822-268
Anti-PPP1R3A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: PPP1R3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR