Anti-TBCA Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A36204-100UG A36204-50UG
77801-929EA 330 USD
77801-929 77801-930
Anti-TBCA Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TBCA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR