Anti-TMEM196 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A36862-100UG A36862-50UG
77811-007EA 330 USD
77811-007 77803-007
Anti-TMEM196 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: TMEM196
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR