Anti-P Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A50555-100UG A50555-50UG
77815-514EA 330 USD
77815-514 77815-515
Anti-P Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hepatitis B virus genotype A2 subtype adw2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR