Anti-ache Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66988-50UG A66988-100UG
77835-998EA 225 USD
77835-998 77835-997
Anti-ache Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ache
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR