Deuterium oxide (99.8% D)

Supplier: TCI America

Synonyms: Water-D2, Heavy water

W0002-10ML W0002-100ML
TCW0002-10ML TCW0002-100ML
Deuterium oxide (99.8% D)
Deuterium oxide

CAS Number: 7789-20-0 MDL Number: MFCD00044636 Molecular Formula: D2O Molecular Weight: 20.03 Form: Clear Liquid Color: Colorless Boiling point (°C): 101 Melting point (°C): 4 Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Water-d2

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.3
Melting Pt: 3.8
Density: 1.11 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6272c81fff9fec28bccb31e813a21fa0d9a0a03a&vid=6272c81fff9fec28bccb31e813a21fa0d9a0a03a&prid=web&clid=SAPDAM

Order Now

</