Wright's stain
Supplier: TCI America
|
Warning
|
W0001-250G
W0001-25G
TCW0001-250GEA
311.8
USD
TCW0001-250G
TCW0001-025G
Wright's stain
Wright's stain
[for Microscopy and Bacteriology]
CAS Number: 68988-92-1
MDL Number: MFCD00082143
Form: Crystal
Color: Deep Green
Flash Point (°C): 11
Density:
0.8 g/cm³ (20 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082143 CAS Number: 68988-92-1 EINECS: 273-541-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...