Triphosgene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Bis(trichloromethyl) carbonate
T1467-250G
T1467-25G
TCT1467-250GEA
595.99
USD
TCT1467-250G
TCT1467-25G
Triphosgene ≥98.0% (by titrimetric analysis)
Triphosgene
CAS Number: 32315-10-9
MDL Number: MFCD00062848
Molecular Formula: C3Cl6O3
Molecular Weight: 296.73
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Phosgene & Thiophosgene Alternatives
Other Product Names / Synonyms: Bis(trichloromethyl) Carbonate
Formula:
C₃Cl₆O₃ MW: 296.75 g/mol Boiling Pt: 206 °C Melting Pt: 80 °C Density: 1.629 g/cm³ (80 °C) |
MDL Number:
MFCD00062848 CAS Number: 32315-10-9 EINECS: 250-986-3 UN: 2928 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...