2-Thiophenemethanol ≥98.0%

Supplier: TCI America

Synonyms: Thiophen-2-ylmethanolThiophen-2-methanol2-Thienylmethanol

T1112-500G T1112-25G T1112-100G
TCT1112-100GEA 432 USD
TCT1112-100G TCT1112-500G TCT1112-025G
2-Thiophenemethanol ≥98.0%
2-Thiophenemethanol

CAS Number: 636-72-6
MDL Number: MFCD00005454
Molecular Formula: C5H6OS
Molecular Weight: 114.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 207
Flash Point (°C): 110
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 2-Hydroxymethylthiophene

Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 207 °C
Melting Pt: 0 °C
Density: 1.21 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3
https://us.vwr.com/stibo/web/std.lang.all/51/78/28995178.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR