(±)-α-Terpineol ≥95.0%

Supplier: TCI America
Warning

Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineolα-Terpineol

T0984-100ML T0984-500ML T0984-25ML
TCT0984-500MLEA 295.07 USD
TCT0984-500ML TCT0984-025ML TCT0984-100ML
(±)-α-Terpineol ≥95.0%
(±)-α-Terpineol

CAS Number: 98-55-5
MDL Number: MFCD00001557
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 218
Flash Point (°C): 101
Freezing point (°C): 31


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 1-p-Menthen-8-ol

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 218 °C
Flash Pt: 101 °C
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
https://us.vwr.com/stibo/web/std.lang.all/50/68/28995068.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR