2,2,6,6-Tetramethylpiperidin-4-ol ≥98.0%

Supplier: TCI America

Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

T0910-25G T0910-500G
TCT0910-025GEA 54.4 USD
TCT0910-025G TCT0910-500G
2,2,6,6-Tetramethylpiperidin-4-ol ≥98.0%
2,2,6,6-Tetramethylpiperidin-4-ol

CAS Number: 2403-88-5 MDL Number: MFCD00005983 Molecular Formula: C9H19NO Molecular Weight: 157.26 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Color: White Melting point (°C): 132 Flash Point (°C): 124


Other Product Names / Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol

Formula: C₉H₁₉NO
MW: 157.26 g/mol
Melting Pt: 132 °C
Flash Pt: 124 °C
MDL Number: MFCD00005983
CAS Number: 2403-88-5
EINECS: 219-291-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=322984127382a062fbddbedd3a1bcf4ffa81c487&vid=322984127382a062fbddbedd3a1bcf4ffa81c487&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR