Tetrachlorosilane ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Silicon (IV) chloride Silicon tetrachloride

T0636-100G T0636-500G T0636-25G
TCT0636-25GEA 26.9 USD
TCT0636-25G TCT0636-100G TCT0636-500G
Tetrachlorosilane ≥98.0% (by titrimetric analysis)
Tetrachlorosilane

CAS Number: 10026-04-7
MDL Number: MFCD00011229
Molecular Formula: SiCl4
Molecular Weight: 169.89
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 59
Melting point (°C): -68
Specific Gravity (20/20): 1.48


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Silicon Tetrachloride

Formula: Cl₄Si
MW: 169.9 g/mol
Boiling Pt: 59 °C
Melting Pt: –68 °C
Density: 1.48 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 12,08644
https://us.vwr.com/stibo/web/std.lang.all/48/26/28994826.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR