2,2,2-Trifluoroethanol ≥99.0%

Supplier: TCI America
Danger

Synonyms: Trifluoro Ethanol, TFE

T0435-25G T0435-500G T0435-100G
TCT0435-025GEA 23.15 USD
TCT0435-025G TCT0435-500G TCT0435-100G
2,2,2-Trifluoroethanol ≥99.0%
2,2,2-Trifluoroethanol

CAS Number: 75-89-8 MDL Number: MFCD00004672 Molecular Formula: C2H3F3O Molecular Weight: 100.04 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 74 Melting point (°C): -44 Flash Point (°C): 30 Specific Gravity (20/20): 1.40


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks Other Product Names / Synonyms: TFE

Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 74.1
Melting Pt: -43.5
Density: 1.40 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 2929
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de69945c97b32866e8f7fbfec3eac0d4d388baf&vid=b72d6e335400f7817bcad126cff4af5e24762dc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR