Trifluoroacetic anhydride ≥98.0%

Supplier: TCI America
Danger

Synonyms: Trifluoracetic anhydrideTFAA

T0433-400ML T0433-100ML T0433-20ML
TCT0433-100MLEA 137.17 USD
TCT0433-100ML TCT0433-400ML TCT0433-20ML
Trifluoroacetic anhydride ≥98.0%
Trifluoroacetic anhydride

CAS Number: 407-25-0
MDL Number: MFCD00000416
Molecular Formula: C4F6O3
Molecular Weight: 210.03
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 40
Specific Gravity (20/20): 1.50


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Fluorinated Building Blocks, Other Fluorinated Group Introducing Agents, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: TFAA

Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 40 °C
Melting Pt: –65 °C
Density: 1.50 (20 °C)
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8,I
https://us.vwr.com/stibo/web/std.lang.all/46/99/28994699.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR