D-(+)-Trehalose dihydrate ≥98.0%

Supplier: TCI America

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

T0331-25G T0331-500G
TCT0331-025GEA 42.84 USD
TCT0331-025G TCT0331-500G
D-(+)-Trehalose dihydrate ≥98.0%
D(+)-Trehalose dihydrate

CAS Number: 6138-23-4 MDL Number: MFCD00071594 Molecular Formula: C12H22O11 Molecular Weight: 342.30 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Specific rotation [a]20/D: 181 deg (C=7, H2O)

Formula: C₁₂H₂₂O₁₁·2H₂O
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d4e54431783e8d387987e004e5ae8cee6b2c2a9&vid=9d4e54431783e8d387987e004e5ae8cee6b2c2a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR