4-Toluoyl chloride ≥98.0%

Supplier: TCI America
Danger

Synonyms: p-Toluoyl chloride, 4-Methylbenzene-1-carbonyl chloride, p-Methylbenzoyl chloride, 4-Methylbenzoyl chloride

T0311-500G T0311-25G T0311-100G
TCT0311-500GEA 114 USD
TCT0311-500G TCT0311-25G TCT0311-100G
4-Toluoyl chloride ≥98.0%
4-Toluoyl chloride

CAS Number: 874-60-2 MDL Number: MFCD00000696 Molecular Formula: C8H7ClO Molecular Weight: 154.59 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 216 Flash Point (°C): 82 Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 4-Methylbenzoyl Chloride

Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 225
Melting Pt: -2
Density: 1.17 (20 °C)
Flash Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00000696
CAS Number: 874-60-2
EINECS: 212-864-8
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33ec410d6cb12dd76273b7e8552242c78a98f42c&vid=33ec410d6cb12dd76273b7e8552242c78a98f42c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR