Tetraethylammonium hydroxide 10% in water

Supplier: TCI America
Danger

T0096-500ML T0096-25ML
TCT0096-500MLEA 96 USD
TCT0096-500ML TCT0096-25ML
Tetraethylammonium hydroxide 10% in water
Tetraethylammonium hydroxide

CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H21NO
Molecular Weight: 147.26
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00

Formula: C₈H₂₁NO
Density: 1.00 (20 °C)
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284000c14d089a1a7a5a4acf51b0b0af8c98b2be&vid=2607ef0c086f77dea88be9f63fa1587517376465&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR