Salicylhydrazide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxybenzene-1-carbohydrazideSalicylic acid hydrazide2-Hydroxybenzoic acid hydrazide2-Hydroxybenzoic hydrazide2-HydroxybenzohydrazideSalicylohydrazide2-Hydroxybenzhydrazide

S0242-25G
TCS0242-025GEA 116.73 USD
TCS0242-025G
Salicylhydrazide ≥98.0% (by HPLC, titration analysis)
Salicylhydrazide

CAS Number: 936-02-7
MDL Number: MFCD00007599
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 150


Other Product Names / Synonyms: Salicylic Hydrazide

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 150 °C
Density: 1.424 g/cm³ (20 °C)
MDL Number: MFCD00007599
CAS Number: 936-02-7
EINECS: 213-311-3
https://us.vwr.com/stibo/web/std.lang.all/40/02/28994002.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR