5-Sulfosalicylic acid dihydrate ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

S0137-500G S0137-25G
TCS0137-500GEA 172 USD
TCS0137-500G TCS0137-25G
5-Sulfosalicylic acid dihydrate ≥99.0% (by HPLC, titration analysis)
5-Sulphosalicylic acid dihydrate

CAS Number: 5965-83-3 MDL Number: MFCD00149540 Molecular Formula: C7H6O6S Molecular Weight: 218.18 Purity/Analysis Method: 99.0% (HPLC,T) Form: Crystal Color: White


Other Product Names / Synonyms: 2-Hydroxy-5-sulfobenzoic Acid

Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Melting Pt: 224 °C
Density: 1.695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b830207c058cba1cebe548786ceec99a3ec1a388&vid=b830207c058cba1cebe548786ceec99a3ec1a388&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR