α-Hydroxy-o-tolyl-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
D(−)-Salicin
CAS Number: 138-52-3
MDL Number: MFCD00006590
Molecular Formula: C13H18O7
Molecular Weight: 286.28
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 200
Specific rotation [a]20/D: -62 deg (C=3, H2O)
Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Crude Medicine Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 2-(Hydroxymethyl)phenyl beta-D-Glucopyranoside
Formula:
C₁₃H₁₈O₇ MW: 286.28 g/mol Melting Pt: 200 °C Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00006590 CAS Number: 138-52-3 EINECS: 205-331-6 Merck Index: 12,08476 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...