Quinaldine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Methylquinoline

Q0005-25G Q0005-500G Q0005-100G
TCQ0005-025GEA 24.42 USD
TCQ0005-025G TCQ0005-500G TCQ0005-100G
Quinaldine ≥98.0%
Quinaldine

CAS Number: 91-63-4 MDL Number: MFCD00006756 Molecular Formula: C10H9N Molecular Weight: 143.19 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 247 Melting point (°C): -2 Flash Point (°C): 110 Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Quinaldine

Formula: C₁₀H₉N
Boiling Pt: 247 °C
Melting Pt: –2 °C
Density: 1.06 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006756
CAS Number: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1601dfa5f08e58b01173c87a58719b05b1cc9792&vid=cc30712267a5d72b3c8cd30c61872c447fa99828&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR