Potassium-tert-butoxide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Potassium tert-butanolate
, Potassium t-butoxide
, Potassium-2-methylpropan-2-olate
, Potassium tert-butylate
P1008-100G
P1008-500G
P1008-25G
TCP1008-500GEA
142.88
USD
TCP1008-500G
TCP1008-25G
TCP1008-100G
Potassium-tert-butoxide ≥97.0% (by titrimetric analysis)
Potassium-tert-butoxide
CAS Number: 865-47-4
MDL Number: MFCD00012162
Molecular Formula: C4H9KO
Molecular Weight: 112.21
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 240
Formula:
C₄H₉KO MW: 112.21 g/mol Melting Pt: 240 °C |
MDL Number:
MFCD00012162 CAS Number: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...