Pentafluorobenzamide 99,0

Supplier: TCI America

Synonyms: 2,3,4,5,6-Pentafluorobenzamide

P0929-25G P0929-5G
TCP0929-25GEA 204 USD
TCP0929-25G TCP0929-005G
Pentafluorobenzamide 99,0
Pentafluorobenzamide

CAS Number: 652-31-3 MDL Number: MFCD00007971 Molecular Formula: C7H2F5NO Molecular Weight: 211.09 Purity/Analysis Method: 99.0% (GC) Form: Crystal Color: White Melting point (°C): 148

Formula: C₇H₂F₅NO
MW: 211.09 g/mol
Boiling Pt: 272
Melting Pt: 151
MDL Number: MFCD00007971
CAS Number: 652-31-3
EINECS: 211-488-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0049ab4b2129ac6b4d258427f1a02d680a1e6dd0&vid=0049ab4b2129ac6b4d258427f1a02d680a1e6dd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR