4-Propylphenol ≥99.0%

Supplier: TCI America

Synonyms: p-Propylphenol, 4-n-Propylphenol

P0698-5G P0698-25G
TCP0698-005GEA 50 USD
TCP0698-005G TCP0698-025G
4-Propylphenol ≥99.0%
4-Propylphenol

CAS Number: 645-56-7 MDL Number: MFCD00002395 Molecular Formula: C9H12O Molecular Weight: 136.19 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 232 Flash Point (°C): 106 Freezing point (°C): 22

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 232
Melting Pt: 24
Density: 1.0091 g/cm³ (0 °C)
Flash Pt: 106 °C
MDL Number: MFCD00002395
CAS Number: 645-56-7
EINECS: 211-446-2
UN: 3145
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a9a59812ead013827987ac85a07e6ca9f1883ad&vid=7284576ea24cfc51e548cb4c7a90dd4c46f6d2f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR