Pyrrolidine ≥98.0%

Supplier: TCI America
Danger

Synonyms: Tetrahydropyrrole

P0576-25ML P0576-500ML
TCP0576-25MLEA 39.91 USD
TCP0576-25ML TCP0576-500ML
Pyrrolidine ≥98.0%
Pyrrolidine

CAS Number: 123-75-1
MDL Number: MFCD00005249
Molecular Formula: C4H9N
Molecular Weight: 71.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Melting point (°C): -63
Flash Point (°C): 3
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Tetramethyleneimine

Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 88 °C
Melting Pt: –63 °C
Density: 0.86 (20 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3,II
Merck Index: 12,08199

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR