Phthalaldehyde ≥99.0%

Supplier: TCI America
Danger

P0280-5G P0280-25G P0280-100G
TCP0280-005GEA 75 USD
TCP0280-005G TCP0280-025G TCP0280-100G
Phthalaldehyde ≥99.0%
Phthalaldehyde

[for HPLC Labeling] CAS Number: 643-79-8 MDL Number: MFCD00003335 Molecular Formula: C8H6O2 Molecular Weight: 134.13 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 84 Melting point (°C): 56 Flash Point (°C): 132


Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents) Other Product Names / Synonyms: Phthaldialdehyde, Benzene-1,2-dicarboxaldehyde

Formula: C₈H₆O₂
Boiling Pt: 84 °C
Melting Pt: 56 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00003335
CAS Number: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4363edc3c76dd8d8d563ba70bd86f7102f83c951&vid=616d154019da07ffbee742061fa3b36b2d91169d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR