Diphenyl ether ≥99.0%
Supplier: TCI America
|
|
Synonyms:
Diphenyl oxide, Phenyl ether, Phenoxybenzene
P0177-500G
P0177-25G
TCP0177-500GEA
41.03
USD
TCP0177-500G
TCP0177-25G
Diphenyl ether ≥99.0%
Diphenyl ether
CAS Number: 101-84-8
MDL Number: MFCD00003034
Molecular Formula: C12H10O
Molecular Weight: 170.21
Purity/Analysis Method:
Other Product Names / Synonyms: Diphenyl Ether
Formula:
C₁₂H₁₀O Boiling Pt: 258 °C Melting Pt: 24…27 °C Density: 1.07 g/cm³ (20 °C) Flash Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003034 CAS Number: 101-84-8 EINECS: 202-981-2 UN: 3077 ADR: 9,III Merck Index: 12,07442 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...