Paraformaldehyde ≥90.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Polyoxymethylene

P0018-500G P0018-25G
TCP0018-500GEA 42.94 USD
TCP0018-500G TCP0018-25G
Paraformaldehyde ≥90.0% (by titrimetric analysis)
Paraformaldehyde

CAS Number: 30525-89-4
MDL Number: MFCD00133991
Molecular Formula: CH2O
Purity/Analysis Method: 90.0% (T)
Form: Crystal
Flash Point (°C): 78

Formula: (CH₂O)n
Boiling Pt: 135…136 °C (13 mmHg)
Melting Pt: 123 °C
Density: 1.46 g/cm³ (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1,III
Merck Index: 12,07158
https://us.vwr.com/stibo/web/std.lang.all/07/41/28990741.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR