Oleic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(9Z)-9-Octadecenoic acid
CAS Number: 112-80-1
MDL Number: MFCD00064242
Molecular Formula: C18H34O2
Molecular Weight: 282.47
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 285
Flash Point (°C): 187
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols, Antioxidants
Other Product Names / Synonyms: cis-9-Octadecenoic Acid
Formula:
C₁₈H₃₄O₂ MW: 282.47 g/mol Boiling Pt: 285 °C Melting Pt: 16.7 °C Density: 0.89 (20 °C) Flash Pt: 187 °C Storage Temperature: Freezer |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 13,06898 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...