Diethyl oxalate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Oxalic acid diethyl ester

O0078-500G O0078-25G
TCO0078-25GEA 22.8 USD
TCO0078-25G TCO0078-500G
Diethyl oxalate ≥98.0%
Diethyl oxalate

CAS Number: 95-92-1
MDL Number: MFCD00009119
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Melting point (°C): -41
Flash Point (°C): 76
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Oxalic Acid Diethyl Ester

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185 °C
Melting Pt: –41 °C
Density: 1.08 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1,III
Merck Index: 13,03152
https://us.vwr.com/stibo/web/std.lang.all/03/87/28990387.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR