Caprylic acid ≥98.0%

Supplier: TCI America
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

O0027-500ML O0027-25ML
TCO0027-500MLEA 58.85 USD
TCO0027-500ML TCO0027-25ML
Caprylic acid ≥98.0%
Caprylic acid

CAS Number: 124-07-2 MDL Number: MFCD00004429 Molecular Formula: C8H16O2 Molecular Weight: 144.21 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 238 Flash Point (°C): 109 Freezing point (°C): 15 Specific Gravity (20/20): 0.91

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7
Melting Pt: 16
Density: 0.91 (20 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8 III
Merck Index: 13,01771
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8baff9f55c6508039e8ff1f7c22673d6be99ea8c&vid=2edb695e0219190de2a1295502164a676b16d239&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR