Nickel(II) chloride, anhydrous ≥98.0%

Supplier: TCI America
Danger

Synonyms: Nickel dichloride

N0850-25G N0850-500G
TCN0850-25GEA 69.84 USD
TCN0850-25G TCN0850-500G
Nickel(II) chloride, anhydrous ≥98.0%
Nickel (II) chloride

CAS Number: 7718-54-9 MDL Number: MFCD00011142 Molecular Formula: Cl2Ni Molecular Weight: 129.59 Purity/Analysis Method: 98.0% (T) Form: Crystal

Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009
Density: 3.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 12,06588

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR