5-Nitrouracil ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Nitrouracil, 5-Nitropyrimidine-2,4(1H,3H)-dione, 2,4-Dihydroxy-5-nitropyrimidine

N0281-25G N0281-5G
TCN0281-025GEA 42.9 USD
TCN0281-025G TCN0281-005G
5-Nitrouracil ≥99.0% (by HPLC, titration analysis)
5-Nitrouracil

CAS Number: 611-08-5 MDL Number: MFCD00006021 Molecular Formula: C4H3N3O4 Molecular Weight: 157.09 Purity/Analysis Method: 99.0% (HPLC,T) Form: Crystal

Formula: C₄H₃N₃O₄
MW: 157.09 g/mol
Melting Pt: 300 °C
Density: 1.807 g/cm³ (20 °C)
MDL Number: MFCD00006021
CAS Number: 611-08-5
EINECS: 210-250-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0642a5bd0622bc1582cb0ce9eab4768be640615f&vid=0642a5bd0622bc1582cb0ce9eab4768be640615f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR