Nicotinamide 99,0

Supplier: TCI America
Warning

Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3

N0078-500G N0078-25G
TCN0078-500GEA 57.5 USD
TCN0078-500G TCN0078-025G
Nicotinamide 99,0
Nicotinamide

CAS Number: 98-92-0 MDL Number: MFCD00006395 Molecular Formula: C6H6N2O Molecular Weight: 122.13 Purity/Analysis Method: 99.0% (HPLC,T) Form: Crystal Melting point (°C): 129 Flash Point (°C): 150


Product Categories: Bioscience, Other Plant Hormones, Reagents for Life-style related Disease Models, Vitamins Other Product Names / Synonyms: Niacinamide

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Melting Pt: 129 °C
Density: 1.4463 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb00153ef341266fdf187f818a35f40ca2f2921&vid=ddb00153ef341266fdf187f818a35f40ca2f2921&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR