1-Naphtholbenzeine
Supplier: TCI America
|
Warning
|
Synonyms:
alpha-Naphtholbenzein
, α-Naphtholbenzein
, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one
, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol
, α-Naphtholbenzeine
, p-Naphtholbenzeine
, 1-Naphtholbenzein
, Bis(4-hydroxy-1-naphthyl)phenylmethanol
, p-Naphtholbenzein
CAS Number: 145-50-6
MDL Number: MFCD00078492
Molecular Formula: C27H18O2
Molecular Weight: 374.44
Form: Crystal
Color: Yellow Red
Melting point (°C): 235
Formula:
C₂₇H₁₈O₂ MW: 374.44 g/mol Melting Pt: 235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078492 CAS Number: 145-50-6 EINECS: 205-656-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...