1-Naphthylacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

N0005-25G N0005-500G
TCN0005-025GEA 38.65 USD
TCN0005-025G TCN0005-500G
1-Naphthylacetic acid ≥98.0% (by titrimetric analysis)
1-Naphthylacetic acid

CAS Number: 86-87-3 MDL Number: MFCD00004046 Molecular Formula: C12H10O2 Molecular Weight: 186.21 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 132


Product Categories: Bioscience, Auxins Other Product Names / Synonyms: 1-Naphthylacetic Acid, NAA

Formula: C₁₂H₁₀O₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 132 °C
Density: ∼1.263 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004046
CAS Number: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a19ce715825eddc954d91493a8ac53e48236b24&vid=2a19ce715825eddc954d91493a8ac53e48236b24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR