2-Tolylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M1313-5G
M1313-25G
TCM1313-005GEA
36
USD
TCM1313-005G
TCM1313-25G
2-Tolylboronic acid (contains varying amounts of Anhydride)
2-Tolylboronic acid
CAS Number: 16419-60-6
MDL Number: MFCD00093526
Molecular Formula: C7H9BO2
Molecular Weight: 135.96
Form: Crystal
Color: White
Other Product Names / Synonyms: 2-Methylbenzeneboronic Acid, o-Tolylboronic Acid
Formula:
C₇H₉BO₂ Melting Pt: 160…166 °C |
MDL Number:
MFCD00093526 CAS Number: 16419-60-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...